Assay Details
| Gene Symbol: Wdr26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145514.5 | NP_663489.4 | 4 - 5 | 1244 | 74 | |
| GenBank mRNA | AK075743.1 | - | 4 - 5 | 636 | 74 | |
| AK082326.1 | - | 5 - 6 | 539 | 74 | ||
| AK205717.1 | - | 3 - 4 | 329 | 74 | ||
| AK214736.1 | - | 3 - 4 | 304 | 74 | ||
| AK219748.1 | - | 4 - 5 | 319 | 74 | ||
| BC058601.1 | - | 2 - 3 | 240 | 74 | ||
| BC094233.1 | - | 1 - 2 | 129 | 74 | ||
| BC139336.1 | - | 4 - 5 | 406 | 8862358 | 74 | |
| BC139337.1 | - | 4 - 5 | 406 | 8862359 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr26
Genomic Map