Assay Details
| Gene Symbol: Iars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198653.2 | NP_941055.1 | 1 - 2 | 383 | 65 | |
| GenBank mRNA | AK008042.1 | - | 1 - 2 | 362 | 65 | |
| AK036182.1 | - | 1 - 2 | 351 | 65 | ||
| AK043590.1 | - | 1 - 2 | 385 | 65 | ||
| AK048653.1 | - | 1 - 2 | 384 | 65 | ||
| AK050358.1 | - | 1 - 2 | 366 | 65 | ||
| AK051778.1 | - | 1 - 2 | 344 | 65 | ||
| AK078183.1 | - | 1 - 2 | 384 | 65 | ||
| AK085714.1 | - | 1 - 2 | 380 | 65 | ||
| AK088965.1 | - | 1 - 2 | 343 | 65 | ||
| AK166832.1 | - | 1 - 2 | 65 | |||
| BC052403.1 | - | 1 - 2 | 360 | 5715723 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Iars2
Genomic Map