Assay Details
| Gene Symbol: Dlg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243047.1 | NP_001229976.1 | 1 - 2 | 248 | 72 | |
| GenBank mRNA | AK039754.1 | - | 1 - 2 | 113 | 72 | |
| CB520190.1 | - | 1 - 2 | 248 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001243047.1 transcripts.
Set Membership: |
|

Dlg2
Genomic Map