Assay Details
| Gene Symbol: Ttll13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177765.3 | NP_808433.2 | 2 - 3 | 562 | 71 | |
| GenBank mRNA | AK029565.1 | - | 2 - 3 | 563 | 71 | |
| AK161519.1 | - | 2 - 3 | 384 | 71 | ||
| AM690757.1 | - | 1 - 2 | 263 | 71 | ||
| BC024578.1 | - | 1 - 2 | 564 | 71 | ||
| BC145749.1 | - | 1 - 2 | 170 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK029565.1 transcripts.
Set Membership: |
|

Ttll13
Genomic Map