Assay Details
Gene Symbol: Igf1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010513.2 | NP_034643.2 | 8 - 9 | 2429 | 85 | |
GenBank mRNA | AF056187.1 | - | 8 - 9 | 1827 | 85 | |
AK137220.1 | - | 3 - 4 | 558 | 85 | ||
AK155884.1 | - | 7 - 8 | 1325 | 85 | ||
BC138868.1 | - | 8 - 9 | 1859 | 8861890 | 85 | |
BC138869.1 | - | 8 - 9 | 1859 | 8861891 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
