Assay Details
| Gene Symbol: Hprt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013556.2 | NP_038584.2 | 3 - 4 | 457 | 125 | |
| GenBank mRNA | AK002286.1 | - | 3 - 4 | 402 | 125 | |
| AK082827.1 | - | 3 - 4 | 347 | 125 | ||
| AK088114.1 | - | 3 - 4 | 347 | 125 | ||
| AK146626.1 | - | 3 - 4 | 403 | 125 | ||
| BC004686.1 | - | 3 - 4 | 372 | 3500885 | 125 | |
| BC083145.1 | - | 3 - 4 | 343 | 30445781 | 125 | |
| J00423.1 | - | 3 - 4 | 398 | 125 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hprt
Genomic Map