Assay Details
| Gene Symbol: Hprt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013556.2 | NP_038584.2 | 7 - 8 | 677 | 86 | |
| GenBank mRNA | AK002286.1 | - | 7 - 8 | 622 | 86 | |
| AK088114.1 | - | 7 - 8 | 567 | 86 | ||
| AK146626.1 | - | 7 - 8 | 623 | 86 | ||
| BC004686.1 | - | 7 - 8 | 592 | 3500885 | 86 | |
| BC083145.1 | - | 7 - 8 | 563 | 30445781 | 86 | |
| J00423.1 | - | 7 - 8 | 618 | 86 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hprt
Genomic Map