Assay Details
| Gene Symbol: Setdb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK089197.1 | - | 4 - 5 | 814 | 77 | |
| BC157926.1 | - | 4 - 5 | 684 | 9088099 | 77 | |
| BC172021.1 | - | 4 - 5 | 684 | 9054307 | 77 | |
| EU155090.1 | - | 4 - 5 | 676 | 77 | ||
| EU155091.1 | - | 4 - 5 | 676 | 77 | ||
| EU155092.1 | - | 4 - 5 | 694 | 77 | ||
| EU155096.1 | - | 3 - 4 | 333 | 77 | ||
| EU155097.1 | - | 5 - 6 | 735 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Setdb2
Genomic Map