Assay Details
| Gene Symbol: Setdb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081024.1 | NP_001074493.1 | 6 - 7 | 955 | 91 | |
| GenBank mRNA | BC157926.1 | - | 5 - 6 | 777 | 9088099 | 91 |
| BC172021.1 | - | 5 - 6 | 777 | 9054307 | 91 | |
| EU155090.1 | - | 5 - 6 | 769 | 91 | ||
| EU155091.1 | - | 5 - 6 | 769 | 91 | ||
| EU155092.1 | - | 5 - 6 | 787 | 91 | ||
| EU155093.1 | - | 4 - 5 | 703 | 91 | ||
| EU155094.1 | - | 4 - 5 | 703 | 91 | ||
| EU155096.1 | - | 4 - 5 | 426 | 91 | ||
| EU155097.1 | - | 6 - 7 | 828 | 91 | ||
| EU155098.1 | - | 4 - 5 | 721 | 91 | ||
| EU155099.1 | - | 5 - 6 | 787 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Setdb2
Genomic Map