Assay Details
| Gene Symbol: Setdb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081024.1 | NP_001074493.1 | 9 - 10 | 1802 | 108 | |
| GenBank mRNA | BC157926.1 | - | 8 - 9 | 1624 | 9088099 | 108 |
| BC172021.1 | - | 8 - 9 | 1624 | 9054307 | 108 | |
| EU155090.1 | - | 8 - 9 | 1616 | 108 | ||
| EU155091.1 | - | 8 - 9 | 1616 | 108 | ||
| EU155092.1 | - | 8 - 9 | 1634 | 108 | ||
| EU155093.1 | - | 7 - 8 | 1550 | 108 | ||
| EU155094.1 | - | 7 - 8 | 1550 | 108 | ||
| EU155096.1 | - | 7 - 8 | 1273 | 108 | ||
| EU155097.1 | - | 9 - 10 | 1675 | 108 | ||
| EU155099.1 | - | 8 - 9 | 1634 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Setdb2
Genomic Map