Assay Details
| Gene Symbol: Mmp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008608.3 | NP_032634.3 | 8 - 9 | 1564 | 74 | |
| GenBank mRNA | AK040968.1 | - | 1537 | 74 | ||
| AK088476.1 | - | 8 - 9 | 1564 | 74 | ||
| AK134340.1 | - | 8 - 9 | 1496 | 74 | ||
| AK138611.1 | - | 8 - 9 | 1505 | 74 | ||
| AK149636.1 | - | 8 - 9 | 1493 | 74 | ||
| AK149907.1 | - | 8 - 9 | 1544 | 74 | ||
| AK149988.1 | - | 8 - 9 | 1464 | 74 | ||
| AK155117.1 | - | 8 - 9 | 1566 | 74 | ||
| AK163601.1 | - | 8 - 9 | 1556 | 74 | ||
| AK165014.1 | - | 8 - 9 | 1565 | 74 | ||
| AK185366.1 | - | 1 - 2 | 112 | 74 | ||
| AK189497.1 | - | 1 - 2 | 112 | 74 | ||
| BC076638.1 | - | 8 - 9 | 1556 | 30658833 | 74 | |
| DQ249870.1 | - | 8 - 9 | 1302 | 74 | ||
| U54984.1 | - | 8 - 9 | 1548 | 74 | ||
| X83536.2 | - | 8 - 9 | 1474 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mmp14
Genomic Map