Assay Details
| Gene Symbol: Slc7a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016972.2 | NP_058668.1 | 1 - 2 | 708 | 80 | |
| GenBank mRNA | AF171668.1 | - | 1 - 2 | 363 | 80 | |
| AK044620.1 | - | 1 - 2 | 769 | 80 | ||
| BC059004.1 | - | 1 - 2 | 708 | 6849059 | 80 | |
| Y19022.1 | - | 1 - 2 | 375 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc7a8
Genomic Map