Assay Details
| Gene Symbol: Lect1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010701.2 | NP_034831.2 | 2 - 3 | 370 | 117 | |
| GenBank mRNA | AK013975.1 | - | 2 - 3 | 284 | 117 | |
| BC045152.1 | - | 2 - 3 | 269 | 6439864 | 117 | |
| U43509.1 | - | 2 - 3 | 371 | 117 | ||
More Information
Set Membership: |
|

Lect1
Genomic Map