Assay Details
| Gene Symbol: Zdhhc20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029492.4 | NP_083768.4 | 4 - 5 | 448 | 81 | |
| GenBank mRNA | AK017158.1 | - | 4 - 5 | 449 | 81 | |
| AK031184.1 | - | 4 - 5 | 407 | 81 | ||
| AK049270.1 | - | 4 - 5 | 428 | 81 | ||
| AK140182.1 | - | 4 - 5 | 409 | 81 | ||
| AK151840.1 | - | 4 - 5 | 463 | 81 | ||
| AK163174.1 | - | 4 - 5 | 446 | 81 | ||
| AK170017.1 | - | 4 - 5 | 466 | 81 | ||
| AK170139.1 | - | 4 - 5 | 389 | 81 | ||
| AY668951.1 | - | 4 - 5 | 366 | 81 | ||
| BC019536.1 | - | 4 - 5 | 465 | 4039734 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zdhhc20
Genomic Map