Assay Details
| Gene Symbol: Chmp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134078.4 | NP_598839.2 | 5 - 6 | 919 | 58 | |
| GenBank mRNA | BB621791.1 | - | 1 - 2 | 52 | 58 | |
| BC006648.1 | - | 1 - 2 | 1123 | 58 | ||
| BC024115.1 | - | 5 - 6 | 907 | 5100379 | 58 | |
| BC033365.1 | - | 5 - 6 | 890 | 5367653 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chmp7
Genomic Map