Assay Details
| Gene Symbol: Pomc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278581.1 | NP_001265510.1 | 2 - 3 | 277 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001278581.1 transcripts.
Set Membership: |
|

Pomc
Genomic Map