Assay Details
| Gene Symbol: Disp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278219.1 | NP_001265148.1 | 2 - 3 | 298 | 116 | |
| GenBank mRNA | AK170113.1 | - | 2 - 3 | 293 | 116 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK170113.1, NM_001278219.1 transcripts.
Set Membership: |
|

Disp1
Genomic Map