Assay Details
Gene Symbol: Larp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138593.2 | NP_613059.2 | 10 - 11 | 1548 | 103 | |
GenBank mRNA | BC029178.1 | - | 2 - 3 | 221 | 103 | |
BC132266.1 | - | 2 - 3 | 219 | 103 | ||
BC132268.1 | - | 2 - 3 | 219 | 103 | ||
BM946683.1 | - | 4 - 5 | 416 | 103 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
