Assay Details
| Gene Symbol: Hmbox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177338.5 | NP_796312.2 | 8 - 9 | 1359 | 79 | |
| GenBank mRNA | AK052729.1 | - | 8 - 9 | 1369 | 79 | |
| AK089782.1 | - | 8 - 9 | 1360 | 79 | ||
| BC002212.1 | - | 6 - 7 | 763 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hmbox1
Genomic Map