Assay Details
| Gene Symbol: Tep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009351.2 | NP_033377.1 | 51 - 52 | 7504 | 153 | |
| GenBank mRNA | AK133745.1 | - | 12 - 13 | 1495 | 153 | |
| AK154212.1 | - | 38 - 39 | 5091 | 153 | ||
| U86137.1 | - | 51 - 52 | 7492 | 153 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tep1
Genomic Map