Assay Details
Gene Symbol: Parp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009632.2 | NP_033762.1 | 14 - 15 | 1394 | 74 | |
GenBank mRNA | AF072521.1 | - | 14 - 15 | 1369 | 74 | |
AJ007780.1 | - | 14 - 15 | 1407 | 74 | ||
BC052000.1 | - | 13 - 14 | 1779 | 74 | ||
BC062150.1 | - | 14 - 15 | 1394 | 6333875 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
