Assay Details
Gene Symbol: Gm6260 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040405.1 | 2 - 3 | 276 | 96 | ||
GenBank mRNA | AK043613.1 | - | 2 - 3 | 278 | 96 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA A830019L24 gene | A830019L24Rik | -412.9kb | Anti-sense | Chr. 3 - 143243055 - 143257996 on NCBI Build GRCm38 | |
Downstream |
predicted gene, 33867 |
Gm33867 | 70.3kb | Anti-sense | Chr. 3 - 143743121 - 143772786 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P972039 | AK043613.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P972039 | AK043613.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 526578 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 424136 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 126840 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 551319 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 29953 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 568495 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 275683 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 973216 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576503 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 445307 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 193129 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 57857 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 95627 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 954506 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 354844 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 162298 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P972039 | AK043613.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P972039 | AK043613.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05102455 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05102476 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05102430 | - | - |
Whole Mouse Genome G4122A | A_52_P972039 | AK043613.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P972039 | AK043613.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122560 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22597998 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598244 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598334 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598338 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02356715 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598124 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598271 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598061 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598069 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598213 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598363 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598023 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02372028 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02883975 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598157 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598345 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598278 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598290 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598315 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598058 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598212 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598098 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598364 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598340 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598007 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598270 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598231 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598326 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598284 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02610490 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02936409 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598229 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03040082 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598165 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598128 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03154734 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598371 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598181 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598303 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03125409 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598321 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598293 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598352 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598305 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598158 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598289 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598060 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598267 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598163 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03094237 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598004 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598312 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598096 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02786331 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02936408 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03078718 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598248 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02672196 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598325 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598175 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598280 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598064 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598361 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02706301 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03050177 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598132 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598261 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598317 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598126 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598243 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02505893 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598272 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02505909 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598122 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598075 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598188 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598034 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598035 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598370 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02441957 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598018 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03078720 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598263 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598265 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598260 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03094239 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598332 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02730794 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598167 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02622340 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598208 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598295 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598350 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598258 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598256 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598277 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02730791 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598194 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598053 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598309 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598005 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598161 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598067 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598329 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598204 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598187 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03078714 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598106 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598070 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598009 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598302 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598020 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598301 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598266 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598247 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02622338 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598185 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598225 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598130 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598115 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598017 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598328 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598268 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598043 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598072 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598039 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598125 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598336 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598029 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598073 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598354 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598276 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598012 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598237 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598211 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598078 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598216 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598037 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598368 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598066 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598346 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03172443 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598169 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598172 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02292504 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598055 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598091 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598015 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598193 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598306 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598054 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03089969 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598358 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598322 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598228 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598372 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598273 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598320 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598025 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598365 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598202 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598286 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598262 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598319 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598341 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02449369 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02601615 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598090 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598205 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598203 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598285 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598207 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02622716 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598215 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02505900 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02505902 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598257 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02672194 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598000 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22597997 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598101 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598235 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03107628 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598206 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598013 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598094 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598246 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02397803 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02695267 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598323 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598347 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02672184 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02954862 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22598349 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR0300024069.mm.1 | NR_040405.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300024069.mm.1 | NR_040405.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300024069.mm.1 | TC0300003061.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300024067.mm.1 | NR_040405.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300024067.mm.1 | NR_040405.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300024067.mm.1 | TC0300003061.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300024068.mm.1 | NR_040405.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300024068.mm.1 | NR_040405.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300024068.mm.1 | TC0300003061.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 535966 | - | - |
