Assay Details
| Gene Symbol: Gm5087 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_121588.1 | 1 - 2 | 126 | 94 | ||
| GenBank mRNA | AK083193.1 | - | 1 - 2 | 127 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK083193.1 transcripts.
Set Membership: |
|

Gm5087
Genomic Map