Assay Details
| Gene Symbol: 1700094M23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK007071.1 | - | 1 - 2 | 148 | 107 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4930482G09 gene | 4930482G09Rik | -23.1kb | Sense | Chr. 3 - 153147943 - 153152108 on NCBI Build GRCm38 | |
| Downstream | ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 3 | St6galnac3 | Siat7c | 19.7kb | Sense | Chr. 3 - 153202509 - 153725133 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

1700094M23Rik
Genomic Map