Assay Details
| Gene Symbol: Gm31739 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK076901.1 | - | 1 - 2 | 428 | 109 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | prostaglandin F receptor | Ptgfr |
AI957154, PGF, fp |
-150.9kb | Anti-sense | Chr. 3 - 151798610 - 151837528 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 31881 |
Gm31881 | 94.0kb | Sense | Chr. 3 - 152084026 - 152107379 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm31739
Genomic Map