Assay Details
Gene Symbol: Cr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007758.2 | NP_031784.1 | 14 - 15 | 2830 | 101 | |
GenBank mRNA | AK003072.2 | - | 14 - 15 | 2830 | 101 | |
BC109344.1 | - | 14 - 15 | 2763 | 40054528 | 101 | |
M29281.1 | - | 9 - 10 | 1834 | 101 | ||
M35684.1 | - | 13 - 14 | 2698 | 101 | ||
M61132.1 | - | 13 - 14 | 2665 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
