Assay Details
| Gene Symbol: 4930455B14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045968.1 | 1 - 2 | 140 | 162 | ||
| GenBank mRNA | AK006409.1 | - | 1 - 2 | 141 | 162 | |
| AK015458.1 | - | 1 - 2 | 85 | 162 | ||
| AK132658.1 | - | 1 - 2 | 141 | 162 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | oncoprotein induced transcript 1 | Oit1 |
2310076N21Rik, AV067083, EF-7, Fam3d |
-287.6kb | Anti-sense | Chr. 14 - 8348937 - 8378763 on NCBI Build GRCm38 |
| Downstream | fragile histidine triad gene | Fhit |
AW045638, Fra14A2 |
883.1kb | Anti-sense | Chr. 14 - 9550092 - 11162054 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4930455B14Rik
Genomic Map