Assay Details
Gene Symbol: Ak5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081277.1 | NP_001074746.1 | 9 - 10 | 1382 | 100 | |
GenBank mRNA | AK053807.1 | - | 9 - 10 | 1382 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
