Assay Details
| Gene Symbol: Cblc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161844.1 | NP_001155316.1 | 9 - 10 | 1380 | 125 | |
| NM_023224.5 | NP_075713.3 | 10 - 11 | 1512 | 125 | ||
| GenBank mRNA | AK010228.1 | - | 10 - 11 | 1513 | 125 | |
| BC046337.1 | - | 10 - 11 | 1488 | 6395806 | 125 | |
| BC111037.1 | - | 9 - 10 | 1364 | 5353413 | 125 | |
| BQ031029.1 | - | 2 - 3 | 125 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Cblc
Genomic Map