Assay Details
Gene Symbol: 4931440J10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045503.1 | 6 - 7 | 528 | 69 | ||
GenBank mRNA | AK016511.1 | - | 6 - 7 | 529 | 69 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | integrator complex subunit 6 | Ints6 |
2900075H24Rik, AI480962, DICE1, Ddx26, HDB, Notch2l |
-67.4kb | Anti-sense | Chr. 14 - 62676325 - 62761370 on NCBI Build GRCm38 |
Downstream | WD repeat and FYVE domain containing 2 | Wdfy2 |
5830485M08, B130024L21Rik, ProF, ZFYVE22 |
7.6kb | Sense | Chr. 14 - 62837690 - 62961509 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 188568 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 78133 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 445976 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 325520 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 421663 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 668898 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 858404 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 17246 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 548922 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 538628 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 910425 | AK002825.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 910425 | AK005487.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 910425 | AK083293.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 910425 | AK089035.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 910425 | AK165299.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 910425 | NM_011515.4 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 910425 | X96737.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 682490 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 755298 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 617760 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 430512 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 444412 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 291152 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 962930 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 558424 | AB207883.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 558424 | AK004802.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 558424 | BC061182.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 558424 | L33412.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 558424 | NM_007425.2 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 147855 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 145017 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 9551 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 453720 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 55857 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 880215 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 171423 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 414052 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 213146 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 213909 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 851098 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 484653 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 176367 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 164699 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 557022 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 33650 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 115599 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 16328 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 9052 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 5478 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 119195 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08135803 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08135870 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02865670 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08135787 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02865695 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02865720 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08135825 | - | - |
Whole Mouse Genome G4122A | A_52_P875166 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132709 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08135825 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29657574 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11208851 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11415464 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11779876 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29657552 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29657565 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29657646 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11430142 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29657554 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11270963 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11203950 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11463833 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29657550 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29657607 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11430141 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29657571 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29657573 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29657564 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29657559 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29657576 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29657606 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29657653 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29657556 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29657553 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29657558 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29657547 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29657605 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29657651 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11463838 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29657647 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29657650 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29657652 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29657603 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29657562 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29657654 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR1400007137.mm.1 | NR_045503.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400007137.mm.1 | NR_045503.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400007137.mm.1 | TC1400000885.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 456426 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 106020563 | - | - |
