Assay Details
Gene Symbol: Gpatch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026367.4 | NP_080643.4 | 5 - 6 | 1214 | 108 | |
GenBank mRNA | AK029990.1 | - | 5 - 6 | 1198 | 108 | |
AK030026.1 | - | 5 - 6 | 1213 | 108 | ||
AK032734.1 | - | 6 - 7 | 1251 | 108 | ||
AK053781.1 | - | 5 - 6 | 1206 | 108 | ||
AK083471.1 | - | 4 - 5 | 371 | 108 | ||
AK169660.1 | - | 5 - 6 | 1215 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
