Assay Details
| Gene Symbol: Oxa1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026936.3 | NP_081212.1 | 4 - 5 | 577 | 129 | |
| GenBank mRNA | AK028379.1 | - | 4 - 5 | 576 | 129 | |
| AK044817.1 | - | 4 - 5 | 576 | 129 | ||
| AK052432.1 | - | 4 - 5 | 578 | 129 | ||
| AK077624.1 | - | 4 - 5 | 578 | 129 | ||
| AK087756.1 | - | 4 - 5 | 581 | 129 | ||
| AK139484.1 | - | 4 - 5 | 585 | 129 | ||
| AK154304.1 | - | 4 - 5 | 580 | 129 | ||
| AK154532.1 | - | 4 - 5 | 578 | 129 | ||
| AK170898.1 | - | 4 - 5 | 578 | 129 | ||
| BC027191.1 | - | 5 - 6 | 509 | 4223847 | 129 | |
More Information
Updated Mapping Information
Set Membership: |
|

Oxa1l
Genomic Map