Assay Details
Gene Symbol: Oxa1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026936.3 | NP_081212.1 | 6 - 7 | 833 | 75 | |
GenBank mRNA | AK007567.1 | - | 6 - 7 | 835 | 75 | |
AK028379.1 | - | 6 - 7 | 832 | 75 | ||
AK044817.1 | - | 6 - 7 | 832 | 75 | ||
AK052432.1 | - | 6 - 7 | 834 | 75 | ||
AK087756.1 | - | 6 - 7 | 837 | 75 | ||
AK139484.1 | - | 6 - 7 | 841 | 75 | ||
AK154304.1 | - | 6 - 7 | 836 | 75 | ||
AK154532.1 | - | 6 - 7 | 834 | 75 | ||
AK170898.1 | - | 6 - 7 | 834 | 75 | ||
BC027191.1 | - | 7 - 8 | 765 | 4223847 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
