Assay Details
| Gene Symbol: Airn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027784.1 | 2 - 3 | 338 | 77 | ||
| GenBank mRNA | BI872620.1 | - | 2 - 3 | 264 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Airn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027784.1 | 2 - 3 | 338 | 77 | ||
| GenBank mRNA | BI872620.1 | - | 2 - 3 | 264 | 77 | |
Set Membership: |
|