Assay Details
| Gene Symbol: 4930447N08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045168.1 | 2 - 3 | 246 | 76 | ||
| GenBank mRNA | AK015409.1 | - | 2 - 3 | 247 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
phosphodiesterase 5A, cGMP-specific |
Pde5a |
Cgbpde, Cn5n, Pde5, Pde5a1 |
Anti-sense | Chr. 3 - 122729158 - 122859374 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 4933405D12 gene | 4933405D12Rik | 82.7kb | Sense | Chr. 3 - 122882592 - 122924116 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4930447N08Rik
Genomic Map