Assay Details
| Gene Symbol: Ceacam20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027839.2 | NP_082115.2 | 5 - 6 | 1060 | 101 | |
| GenBank mRNA | AK018613.1 | - | 5 - 6 | 1057 | 101 | |
| AK033581.2 | - | 5 - 6 | 1061 | 101 | ||
| BC049361.1 | - | 5 - 6 | 1041 | 6476373 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ceacam20
Genomic Map