Assay Details
| Gene Symbol: Ppa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293641.1 | NP_001280570.1 | 1 - 2 | 167 | 120 | |
| NM_146141.2 | NP_666253.1 | 1 - 2 | 167 | 120 | ||
| GenBank mRNA | AA444705.1 | - | 1 - 2 | 166 | 120 | |
| AK090384.1 | - | 1 - 2 | 156 | 120 | ||
| AK143475.1 | - | 1 - 2 | 138 | 120 | ||
| BC011417.1 | - | 1 - 2 | 161 | 2654305 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppa2
Genomic Map