Assay Details
| Gene Symbol: Rcor3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290278.1 | NP_001277207.1 | 5 - 6 | 654 | 124 | |
| NM_001290279.1 | NP_001277208.1 | 5 - 6 | 654 | 124 | ||
| NM_001290280.1 | NP_001277209.1 | 5 - 6 | 654 | 124 | ||
| NM_001290282.1 | NP_001277211.1 | 6 - 7 | 768 | 124 | ||
| NM_144814.3 | NP_659063.3 | 5 - 6 | 654 | 124 | ||
| GenBank mRNA | AK040839.1 | - | 5 - 6 | 641 | 124 | |
| AK050287.1 | - | 6 - 7 | 759 | 124 | ||
| AK135612.1 | - | 5 - 6 | 627 | 124 | ||
| AK155440.1 | - | 5 - 6 | 626 | 124 | ||
| AK193623.1 | - | 1 - 2 | 88 | 124 | ||
| BC020065.1 | - | 5 - 6 | 541 | 124 | ||
| BC057141.1 | - | 5 - 6 | 401 | 5151580 | 124 | |
| BC138938.1 | - | 5 - 6 | 374 | 8861960 | 124 | |
| BC138939.1 | - | 5 - 6 | 374 | 8861961 | 124 | |
| BC145469.1 | - | 5 - 6 | 374 | 124 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rcor3
Genomic Map