Assay Details
| Gene Symbol: Rcor3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290278.1 | NP_001277207.1 | 9 - 10 | 1150 | 75 | |
| NM_001290280.1 | NP_001277209.1 | 9 - 10 | 1150 | 75 | ||
| GenBank mRNA | AK135612.1 | - | 9 - 10 | 1123 | 75 | |
| AK155440.1 | - | 9 - 10 | 1122 | 75 | ||
| BC020065.1 | - | 9 - 10 | 1036 | 75 | ||
| BC145469.1 | - | 9 - 10 | 870 | 75 | ||
More Information
Set Membership: |
|

Rcor3
Genomic Map