Assay Details
Gene Symbol: Larp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138593.2 | NP_613059.2 | 1 - 2 | 161 | 106 | |
GenBank mRNA | AK012772.1 | - | 1 - 2 | 151 | 106 | |
AK015041.1 | - | 1 - 2 | 136 | 106 | ||
AK082815.1 | - | 1 - 2 | 160 | 106 | ||
AK162147.1 | - | 1 - 2 | 78 | 106 | ||
BC023165.1 | - | 1 - 2 | 115 | 106 | ||
BY269910.1 | - | 1 - 2 | 161 | 106 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK012772.1, AK015041.1, AK162147.1, BC023165.1, NM_138593.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
