Assay Details
| Gene Symbol: Lpgat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134829.1 | NP_001128301.1 | 3 - 4 | 580 | 83 | |
| NM_172266.3 | NP_758470.1 | 3 - 4 | 559 | 83 | ||
| GenBank mRNA | AK133814.1 | - | 3 - 4 | 560 | 83 | |
| AK172914.1 | - | 3 - 4 | 530 | 83 | ||
| BC013667.1 | - | 3 - 4 | 468 | 4020003 | 83 | |
| BC024750.1 | - | 1 - 2 | 50 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lpgat1
Genomic Map