Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300022728.mm.1 |
NM_080451.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022728.mm.1 |
NM_080451.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022728.mm.1 |
TC0300002872.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300022737.mm.1 |
NM_080451.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022737.mm.1 |
NM_080451.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022737.mm.1 |
TC0300002872.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300022730.mm.1 |
NM_080451.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022730.mm.1 |
NM_080451.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022730.mm.1 |
TC0300002872.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300022740.mm.1 |
NM_080451.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022740.mm.1 |
NM_080451.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022740.mm.1 |
TC0300002872.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300022739.mm.1 |
NM_080451.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022739.mm.1 |
NM_080451.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022739.mm.1 |
TC0300002872.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300022731.mm.1 |
NM_080451.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022731.mm.1 |
NM_080451.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022731.mm.1 |
TC0300002872.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300022732.mm.1 |
NM_080451.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022732.mm.1 |
NM_080451.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022732.mm.1 |
TC0300002872.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300022729.mm.1 |
NM_080451.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022729.mm.1 |
NM_080451.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022729.mm.1 |
TC0300002872.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300022738.mm.1 |
NM_080451.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022738.mm.1 |
NM_080451.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022738.mm.1 |
TC0300002872.mm.1 |
MTA TC |