Assay Details
| Gene Symbol: Anpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008486.2 | NP_032512.2 | 17 - 18 | 2628 | 77 | |
| GenBank mRNA | AK143746.1 | - | 17 - 18 | 2571 | 77 | |
| AK171427.1 | - | 18 - 19 | 2990 | 77 | ||
| BC005431.1 | - | 17 - 18 | 2594 | 3593307 | 77 | |
| BC017011.1 | - | 17 - 18 | 2652 | 3590117 | 77 | |
| BC040792.1 | - | 17 - 18 | 2652 | 3590117 | 77 | |
| U77083.1 | - | 17 - 18 | 2560 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Anpep
Genomic Map