Assay Details
| Gene Symbol: Fezf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080433.3 | NP_536681.2 | 2 - 3 | 1164 | 82 | |
| GenBank mRNA | AB042399.1 | - | 1 - 2 | 881 | 82 | |
| AK019996.1 | - | 2 - 3 | 1128 | 82 | ||
| BC055718.1 | - | 1 - 2 | 1491 | 6415359 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fezf2
Genomic Map