Assay Details
| Gene Symbol: Zranb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017381.2 | NP_059077.1 | 2 - 3 | 128 | 65 | |
| GenBank mRNA | AF133818.1 | - | 2 - 3 | 106 | 65 | |
| AK146275.1 | - | 2 - 3 | 126 | 65 | ||
| AK146854.1 | - | 2 - 3 | 120 | 65 | ||
| AK150291.1 | - | 2 - 3 | 167 | 65 | ||
| AK152536.1 | - | 2 - 3 | 167 | 65 | ||
| AK166534.1 | - | 2 - 3 | 123 | 65 | ||
| AK167756.1 | - | 2 - 3 | 126 | 65 | ||
| AK167766.1 | - | 2 - 3 | 156 | 65 | ||
| AK167862.1 | - | 2 - 3 | 156 | 65 | ||
| AK167983.1 | - | 2 - 3 | 156 | 65 | ||
| AK168033.1 | - | 2 - 3 | 126 | 65 | ||
| AK168162.1 | - | 2 - 3 | 127 | 65 | ||
| AK168613.1 | - | 2 - 3 | 158 | 65 | ||
| AK169114.1 | - | 2 - 3 | 120 | 65 | ||
| AK169335.1 | - | 2 - 3 | 129 | 65 | ||
| BC049115.1 | - | 2 - 3 | 138 | 65 | ||
| BC132547.1 | - | 2 - 3 | 125 | 40130824 | 65 | |
| BC138575.1 | - | 2 - 3 | 125 | 8861595 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zranb2
Genomic Map