Assay Details
| Gene Symbol: Clca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017474.2 | NP_059502.1 | 10 - 11 | 1719 | 70 | |
| GenBank mRNA | AB017156.1 | - | 10 - 11 | 1702 | 70 | |
| AK007466.1 | - | 10 - 11 | 1717 | 70 | ||
| AK008659.2 | - | 10 - 11 | 1716 | 70 | ||
| AK033560.1 | - | 1721 | 70 | |||
| AK136476.1 | - | 10 - 11 | 1716 | 70 | ||
| BC028343.1 | - | 10 - 11 | 1709 | 4217548 | 70 | |
| BC116318.2 | - | 10 - 11 | 1702 | 40095991 | 70 | |
| BC116319.2 | - | 10 - 11 | 1702 | 40095992 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clca1
Genomic Map