Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300024136.mm.1 |
NM_011828.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024136.mm.1 |
NM_011828.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024136.mm.1 |
TC0300003071.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300024137.mm.1 |
NM_011828.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024137.mm.1 |
NM_011828.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024137.mm.1 |
TC0300003071.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300024138.mm.1 |
NM_011828.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024138.mm.1 |
NM_011828.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024138.mm.1 |
TC0300003071.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300024133.mm.1 |
NM_011828.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024133.mm.1 |
NM_011828.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024133.mm.1 |
TC0300003071.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300024135.mm.1 |
NM_011828.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024135.mm.1 |
NM_011828.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024135.mm.1 |
TC0300003071.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300024139.mm.1 |
NM_011828.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024139.mm.1 |
NM_011828.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024139.mm.1 |
TC0300003071.mm.1 |
MTA TC |