Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100029824.mm.1 |
NM_007422.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100029824.mm.1 |
NM_007422.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100029824.mm.1 |
TC0100003639.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100029826.mm.1 |
NM_007422.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100029826.mm.1 |
NM_007422.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100029826.mm.1 |
TC0100003639.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100029825.mm.1 |
NM_007422.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100029825.mm.1 |
NM_007422.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100029825.mm.1 |
TC0100003639.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100029827.mm.1 |
NM_007422.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100029827.mm.1 |
NM_007422.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100029827.mm.1 |
TC0100003639.mm.1 |
MTA TC |