Assay Details
| Gene Symbol: Gipc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016867.1 | NP_058563.1 | 4 - 5 | 725 | 64 | |
| GenBank mRNA | AF061262.1 | - | 4 - 5 | 725 | 64 | |
| AK008307.1 | - | 4 - 5 | 542 | 64 | ||
| AK008626.1 | - | 4 - 5 | 738 | 64 | ||
| AK168560.1 | - | 4 - 5 | 736 | 64 | ||
| BC013440.1 | - | 4 - 5 | 779 | 4238716 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gipc2
Genomic Map