Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300024639.mm.1 |
NM_027287.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024639.mm.1 |
TC0300003139.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300024638.mm.1 |
NM_027287.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024638.mm.1 |
TC0300003139.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300024626.mm.1 |
NM_025926.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024626.mm.1 |
NM_027287.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024626.mm.1 |
TC0300003139.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300024640.mm.1 |
NM_027287.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300024640.mm.1 |
TC0300003139.mm.1 |
MTA TC |